Would you like to log in again?
IGF-I ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
E20
102994-306EA 921.92 USD InStock
102994-306
IGF-I ELISA , Eagle Biosciences
Assays ELISAs
Human Insulin-like Growth Factor-1 (IGF-1) ELISA determines IGF-1 in serum
  • Dynamic Range: 2 - 50 ng/ml
  • Sensitivity: 0.11 ng/mL
  • Incubation: 1.75 hours
Insulin-like growth factors (IGF) -I and -II play a pivotal role in regulating the proliferation, differentiation and specific functions of many cell types (1-3). IGF-I is identical with Somatomedin C (Sm-C) (4) and has a molecular weight of 7649 daltons (5). Its major regulators are growth hormone (GH) and nutrition (6), although its production in specific tissues is affected by a multitude of tropic hormones and other peptide growth factors. In contrast to many other peptide hormones, IGFs are avidly bound to specific binding proteins (IGFBP). The seven classes of IGFBPs which are known at present (7,8,22) either bind IGF-I and IGF-II with similar affinities or show a preference for IGF-II (9,10).A major problem of IGF-I measurement results from the interference of IGFBPs in the assay. Direct determinations in untreated serum samples (11) give false values because of the extremely slow dissociation of the IGF-I/IGFBP-3 complexes during the assay incubation. Depending on the ratio IGF-I to IGFBP the following errors may occur.
For Research Use Only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty