Would you like to log in again?
Mouse- / Rat-IGF-I ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E25
102994-308EA 810.77 USD InStock
102994-308
Mouse- / Rat-IGF-I ELISA, Eagle Biosciences
Assays ELISAs
Mouse/Rat IGF-I ELISA is to be used for the measurement of IGF-I in mouse/rat serum and plasma.
  • Dynamic Range: 0.5 - 18 ng/mL
  • Sensitivity: 0.029 ng/ml
  • Incubation: 2 hours
Beside different cell culture models and studies with human patients, mice and rats are suitable model organisms for basic research and pre-clinical studies.Thus, we developed this test system as a tool for IGF-I measurements in mice and rat for usage in research and pre-clinical studies. Even if the comparability of mice and humans is limited we offer some background information on the human IGF-I system in the following section:
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty