Would you like to log in again?
ALS (Acid Labile Subunit) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E35
102994-312EA 921.92 USD InStock
102994-312
ALS (Acid Labile Subunit) ELISA, Eagle Biosciences
Assays ELISAs
ALS ELISA is suited for the measuring ALS in human serum or EDTA-/heparin-/citrate plasma.
  • Dynamic Range: 1.5 - 40 mU/ml
  • Sensitivity: 0.2 mU/ml
  • Incubation: 3 hours
The acid-labile Subunit, is a synthezised as propeptide of 605 amino acids. The signal peptide, necessary for ALS secretion (AA 1-27) cleaved off enduring the transport process (Swiss-Prot P35858 Version 82). The mature protein consists of 578 amino acids and contains about 20 leucin rich sequence repeats. Beside the leucin-rich repeats several potential N-linked glycosylation sides have been described. Miller BS et al. were able to demonstrate that incomplete glycolsylation of IGFs, ALS and IGFBP-3 results in a decreased serum concentration of these proteins. Oral mannose therapy resulted in a partial normalization of the glycosylation pattern and went along with improved growth [8]. Mutations in or the complete knock out of the ALS gene result in IGF / IGFBP-3 deficiency and therewith in disturbation of growth [9,10]. Beside growth also other Endocrinology axes may be involved. In primary ALS deficiency hyperinsulinemia could be observed [11, 12]. Further, the ALS-IGF-IGFBP-system seems to be of relevance in coronary disease [13].
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty