Would you like to log in again?
Eosinophil Derived Neurotoxin (EDN) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
EDN39-K01
102994-318EA 1673.85 USD InStock
102994-318
Eosinophil Derived Neurotoxin (EDN) ELISA, Eagle Biosciences
Assays ELISAs
ELISA is intended to detect eosinophil derived neurotoxin (EDN) in stool, plasma, serum and urine.
  • Dynamic Range: 0 - 35 ng/ml
  • Sensitivity: 0.62 ng/ml
  • Incubation: 2 hours
After activation eosinophil granulocytes segregate the cationic glycoprotein EDN (eosinophil derived neurotoxin). This 18-21 kDa single stranded glycosylated protein is also known as EPX (eosinophil protein X). Together with ECP (eosinophil cationic protein), EDN belongs to the ribonuclease superfamily1-3. EDN, however, has a 100-fold increased ribonuclease activity. EDN is also neurotoxic and not cytotoxic4,5. The activation of eosinophil granulocytes is important during the inflammatory processes in allergic reactions. Thus EDN is a marker for eosinophil activation and degranulation. The measurement of EDN in stool allows the detection of clinical or subclinical chronic inflammation in the gut. Some research has suggested that the measurement of EDN gives information on the activity of disease and the prediction of a relapse. The Eosinophil Derived Neurotoxin (EDN) ELISA Kit allows an easy, rapid and precise quantitative determination of the eosinophil derived neurotoxin in biological samples. The kit includes all reagents ready to use for preparation of the samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty