Would you like to log in again?
Anti Endothelin Receptor A IgG3 Antibody ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
EG331-K01
102994-324EA 2720 USD InStock
102994-324
Anti Endothelin Receptor A IgG3 Antibody ELISA, Eagle Biosciences
Assays ELISAs
Anti Endo Receptor A IgG3 Antibody EIA detects antibodies against the Endothelin receptor subtype A.
  • Dynamic Range: 2.5 - 40 U/ml
  • Sensitivity: 2.5 U/mL
  • Incubation: 3.5 hours
Endothelins (ET) are 21-amino acid vasoconstricting peptides produced primarily in the endothelium having a key role in vascular homeostasis. It mediates the effects through G-Protein-coupled receptors, the Endothelin receptors. There are two key receptor types, ETA and ETB. ETA receptors are found in the smooth muscle tissue of blood vessels where they increase vasoconstriction by ET-1.
The Eagle Biosciences Human Anti Endothelin Receptor A IgG3 Antibody is designed for the determination of antibodies (IgG3) against the Endothelin receptor subtype A in serum and plasma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty