Would you like to log in again?
Anti Endothelin Receptor A IgG4 Antibody ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
EG431-K01
102994-326EA 2720 USD InStock
102994-326
Anti Endothelin Receptor A IgG4 Antibody ELISA, Eagle Biosciences
Assays ELISAs
Human Anti Endothelin Receptor A IgG4 Antibody ELISA determines IgG4 antibodies in serum or plasma.
  • Dynamic Range: 2.5 - 40 U/ml
  • Sensitivity: 2.5 U/mL
  • Incubation: 3.5 hours
The Eagle Biosciences Human Anti Endothelin Receptor A IgG4 Antibody ELISA Assay Kit is an antibody screening test. Endothelin- receptor A has been pre-coated onto a microtiter plate. During the first incubation the anti-Endothelin receptor A-Antibodies of the samples are immobilized on the plate. The autoantibodies are detected with a POD labelled anti-human IgG4 antibody. In the following enzymatic substrate reaction the intensity of the color correlates with the concentration and/ or avidity of anti-Endothelin receptor A-antibody.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty