Would you like to log in again?
Human EGF ELISA KIT, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
EGF31-K01
102994-328EA 660.38 USD InStock
102994-328
Human EGF ELISA KIT, Eagle Biosciences, Inc.
Assays ELISAs
Human EGF ELISA detects human Epidermal Growth Factor (EGF) concentrations in various sample types.
  • Dynamic Range: 62.5 - 2000 pg/ml
  • Sensitivity: 15 pg/mL
  • Incubation: 3.5 hours
The Eagle Biosciences Human Epidermal Growth Factor (EGF) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Epidermal Growth Factor (EGF) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Human Epidermal Growth Factor (EGF) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty