Would you like to log in again?
ENA screen (Extractable Nuclear Antigens) ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
4011
102994-330EA 568.85 USD InStock
102994-330
ENA screen (Extractable Nuclear Antigens) ELISA, Eagle Biosciences, Inc.
Assays ELISAs
ENA screen ELISA detects autoantibodies to nuclear and cytoplasmic antigens in human serum, plasma.
  • Dynamic Range: n/a
  • Sensitivity: Cut Off Control
  • Incubation: 2 hours
Systemic autoimmune diseases such as systemic lupus erythematosus, scleroderma, rheumatoid arthritis, Sjögren’s syndrome, dermatomyositis, mixed connective tissue disease are characterized by the appearance of a variety of autoantibodies directed against components of the cell nucleus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty