Would you like to log in again?
Fetuin A ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
FET39-K01
102994-386EA 1144.23 USD InStock
102994-386
Fetuin A ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Fetuin A ELISA detects human Fetuin-A, in serum, plasma, cell culture supernatant, tissue and urine.
  • Dynamic Range: 12.5 - 370 ng/ml
  • Sensitivity: 5.0 ng/mL
  • Incubation: 3 hours
Fetuin-A, also known as alpha-2-HS glycoprotein, is a 59 kDa glycoprotein that consists of two amino-terminal cystatin domains and a smaller carboxyl-terminal domain. Fetuin-A is synthesized by the liver and secreted into blood stream, where its concentration in adult mammals ranges from 0.5 – 1.0 g/L. Fetuin-A occurs in high serum concentration during fetal life and involves in protease inhibitory activities and development-associated regulation of calcium metabolism and osteogenesis. It accumulates in bones and teeth as a major fraction of noncollagenous bone proteins.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty