Would you like to log in again?
Anti-Gliadin sIgA / IgA ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
GLI35-K01
102994-416EA 1144.23 USD InStock
102994-416
Anti-Gliadin sIgA / IgA ELISA, Eagle Biosciences
Assays ELISAs
Anti-Gliadin sIgA / IgA ELISA is intended to detect the Anti-Gliadin sIgA / IgA in stool.
  • Dynamic Range: 25 - 200 mU/g
  • Sensitivity: 1.3 mU/g
  • Incubation: 2 hours
Celiac disease is a chronic illness of the small intestinal mucous membrane. It is caused by an intolerance against gluten, which is found in many cereals. The intake of gluten-containing food leads to inflammation of the small intestinal mucous membrane. The resorption of nutrients is therefore reduced. The symptoms of the disease are reduction of weight, diarrhoea, vomiting, anorexia and tiredness. The growth in children is reduced. The only therapeutic treatment is a gluten-free diet. An untreated celiac disease has been known to increase the risk of non-Hodgkin-lymphoma and colon cancer. In five to ten percent of the patients, celiac disease is associated with diabetes mellitus type 1. Women are more often affected than men. The outcome of the disease is pronounced during infancy and in an age between 30 and 40 years. The Eagle Biosciences Anti-Gliadin sIgA / IgA ELISA kit allows for an easy, rapid and precise quantitative determination of the secretory IgA and IgA gliadin antibodies in stool. The kit includes all reagents ready to use for preparation of the samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty