Would you like to log in again?
Glutathione HPLC Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
GLU31-H100
102994-418EA 1235.77 USD InStock
102994-418
Glutathione HPLC Assay, Eagle Biosciences, Inc.
Assays Immunological Assays
The Glutathione HPLC Assayis for the quantitative determination of glutathione in EDTA blood.
  • Incubation Time: 40 minutes
  • Sample Size: 100 µl
  • Sample Type: EDTA Whole Blood
Most of the intracellular glutathione is reduced (approx. 90 % in EDTA-blood), only 10 % is oxidized (GSSG). The NADPH-dependent glutathione reductase maintains this steady state. Alterations in the glutathione status are involved in the pathogenesis of several diseases. In discussion are reperfusion damage, liver injury, cancer, diabetes mellitus, cataract, inflammatory diseases, chronic lymphatic edema and radiation damages. Altered glutathione concentrations might also be due to pollution, cigarette smoke, side effects of drugs and aging. In case of oxidative stress the amount of reduced glutathione is diminished. The relation of reduced to oxidized glutathione gives information about the redox and detoxification status of cells and tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty