Would you like to log in again?
Mouse / Rat Active GLP-1 ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
GP121-K01
102994-422EA 961.15 USD InStock
102994-422
Mouse / Rat Active GLP-1 ELISA , Eagle Biosciences
Assays ELISAs
Mouse Rat GLP-1 Active (7-36) ELISA detects bioactive GLPe-1 (7-36) amide level in plasma samples.
  • Dynamic Range: .85 - 146 pM
  • Sensitivity: 0.1 pM
  • Incubation: 24.5 hours
The Eagle Biosciences Mouse/Rat Active GLP-1 ELISA Assay Kit (enzyme-linked immunoassay kit) is intended quantitative determination of bioactive glucagon-like peptide-1 (7-36) amide [GLP-1 (7-36)] level in rat and mouse plasma samples with only 20 ul of sample volume. E The primary amino acid sequence of GLP-1 peptide is identical among mammalian species, i.e. rat, mouse, pig, human, etc. Eagle Biosciences Mouse/Rat Active GLP-1 ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty