Would you like to log in again?
Human Granzyme B ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
GRB31-K01
102994-426EA 830.38 USD InStock
102994-426
Human Granzyme B ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
CD44 ELISA detects soluble CD44 in human serum, plasma, buffered solutions or cell culture medium.
  • Dynamic Range: 31.25-1000 pg/ml
  • Sensitivity: 20 pg/ml
  • Incubation: 3h00
The Eagle Biosciences Human Granzyme B ELISA is to be used for the in-vitro quantitative determination of Granzyme B in human serum, plasma, buffered solutions or cell culture medium. The Eagle Biosciences Human Granzyme B ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty