Would you like to log in again?
GSH/GSSG Cuvette Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
GSH39-K01
102994-434EA 699.62 USD InStock
102994-434
GSH/GSSG Cuvette Assay, Eagle Biosciences, Inc.
Assays ELISAs
GSH GSSG Cuvette is to detect the GSH GSSG ratio in biological fluids by a cuvette .
  • Dynamic Range: 0.1 - 3.0 µM
  • Sensitivity: 0.1 µM
  • Incubation: 20 minutes
Reduced glutathione (GSH) is a tripeptide (g-glutamylcysteinylglycine) that contains a free thiol group. GSH is a major tissue antioxidant that provides reducing equivalents for the glutathione peroxidase (GPx) catalyzed reduction of lipid hydroperoxides to their corresponding alcohols and hydrogen peroxide to water. In the GPx catalyzed reaction, the formation of a disulfide bond between two GSH molecules gives rise to oxidized glutathione (GSSG). The enzyme glutathione reductase (GR) recycles GSSG to GSH with the simultaneous oxidation of -nicotinamide adenine dinucleotide phosphate (-NADPH2). When cells are exposed to increased levels of oxidative stress, GSSG will accumulate and the ratio of GSH to GSSG will decrease. Therefore, the determination of the GSH/GSSG ratio and the quantification of GSSG are useful indicators of oxidative stress in cells and tissues.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty