Would you like to log in again?
Human Anti-Mouse Antibody (HAMA) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
HAM31-K01
102994-448EA 843.46 USD InStock
102994-448
Human Anti-Mouse Antibody (HAMA) ELISA, Eagle Biosciences
Assays ELISAs
Human Anti-Mouse Antibody ELISA is intended to detect human Anti-Mouse Antibody levels in serum.
  • Dynamic Range: 40 - 1500 ng/ml
  • Sensitivity: 2 ng/ml
  • Incubation: 2 hours
The Eagle Biosciences Human Anti-Mouse Antibody (HAMA) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Anti-Mouse Antibody (HAMA) levels in serum. The Eagle Biosciences Human Anti-Mouse Antibody (HAMA) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty