Would you like to log in again?
Homocysteine HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
HCY31-H100
102994-450EA 935 USD InStock
102994-450
Homocysteine HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Homocysteine HPLC Assay is intended to determine homocysteine in plasma,serum.
  • Incubation Time: 10 minutes
  • Sample Size: 50 µl
  • Sample Type: Serum, Plasma
Homocysteine is an amino acid, which is produced out of methionine. It is not ingested by nutrition. Homocysteine is rapidly degraded to cysteine when vitamin B6, folic acid and vitamin B12 are sufficiently available. Increased levels of homocysteine will damage heart and blood vessels. Meanwhile it is an established risk marker for cardiovascular disease. US studies demonstrate that 40% of heart attacks are caused by increased homocysteine levels. Also the risk for stroke is increased by elevated homocysteine concentrations in plasma. A participation of homocysteine in Alzheimer and Parkinson diseases are under discussion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty