Would you like to log in again?
8-hydroxy-2’-deoxyguanosine (8-OHdG) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
HDG39-K01
102994-452EA 2059.62 USD InStock
102994-452
8-hydroxy-2’-deoxyguanosine (8-OHdG) ELISA, Eagle Biosciences
Assays ELISAs
8-hydroxy-2'-deoxyguanosine (8-OHdG) EIA detects adduct 8-OHdG in urine,serum or biological samples.
  • Dynamic Range: 0.5 - 200 ng/mL
  • Sensitivity: 0.5 ng/mL
  • Incubation: 3.5 hours
The Eagle Biosciences 8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA assay kit is intended for the quantitative determination of adduct 8-hydroxy-2’-deoxy-guanosine (8-OHdG) in urine, serum or biological samples by enzyme linked immunoassay (ELISA). The 8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA assay kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty