Would you like to log in again?
Helicobacter Antigens ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
HEL31-K01
102994-456EA 542.69 USD InStock
102994-456
Helicobacter Antigens ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Helicobacter pylori Antigen ELISA is to detect Helicobacter pylori antigen in fecal specimen.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2 hours
Helicobacter pylori (H. pylori) is a gram-negative spiral shaped microaerophilic bacterium that can colonize the human stomach. In the stomach H. pylori protects itself from destruction by gastric acid through incorporation into the gastric mucosa and through cleavage of urea to ammonia and carbon dioxide. The ammonia neutralizes the gastric acid in the contiguity of the germ. This reaction is catalyzed by the bacterial enzyme urease that can also be used as diagnostic proof (Helicobacter-Urease-test). H. pylori infections are often associated with stomach disorders characterized by increased production of gastric acid, for example type B gastritis, about 75 % of Ulcus ventriculi and almost all cases of Ulcus duodeni. Therefore the screening of H.pylori infection is an important part in the  diagnostics  of stomach diseases Chronic infections with H. pylori are furthermore an accepted high risk factor for the development of gastric malignomas and MALT lymphoma. Therefore, in 1944 the WHO has dedicated H. pylori into group I of the defined carcinogens.  Immunological tests on the basis of specific anti-H. pylori antibodies enable a noninvasive and direct detection of H. pylori Antigenss from stool specimen and other matrices and may be used for diagnostic screening and for therapeutic surveillance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty