Would you like to log in again?
Hexanoyl-Lys adduct (HEL) ELISA , Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
HEL39-K01
102994-458EA 2059.62 USD InStock
102994-458
Hexanoyl-Lys adduct (HEL) ELISA , Eagle Biosciences, Inc.
Assays ELISAs
The Hexanoyl-Lys adduct (HEL) EIA is intended to determine HEL adduct in biological samples.
  • Dynamic Range: 2.6 - 624 nmol/L
  • Sensitivity: 2.6 nmol/L
  • Incubation: Overnight
Hexanoyl-Lys adduct (HEL) is formed by the reaction of linoleic acid hydroperoxide and Lysine, and a biomarker for oxidative stress. The Hexanoyl­Lysine adduct ELISA assay kit is a competitive enzyme-linked immunosorbent assay for quantitative measurement of hexanoyl-Lys adduct. This kit is based on the monoclonal antibody clone 5H4, which is specific for HEL. Suitable for urine, serum and other biological samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty