Would you like to log in again?
Histamine ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
EA213/96
102994-462EA 575.38 USD InStock
102994-462
Histamine ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Histamine ELISA is intended to detect histamine in Plasma, Urine, and Cell Culture.
  • Dynamic Range: 0.15 - 25 ng/mL
  • Sensitivity: 0.02 ng/mL
  • Incubation: 2.5 hours
Histamine plays a major role in the initial phase of an anaphylactic reaction. The quantification of histamine in plasma after allergen administration is of clinical interest. Histamine is part of the immune response to foreign pathogens and it increases the permeability of the capillaries to white blood cells and other proteins in order to allow them to engage foreign invaders in the affected tissues. Responsible for the biological effects of histamine in tissue are the activation of different surface receptors, for instance H1, H2 and H3. Histamine is involved in the regulating physiological function in the gut and acting as a neurotransmitter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty