Would you like to log in again?
H pylori Antigens ELISA Assay Kit (Quantitative), Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
HPY35-K01
102994-468EA 765 USD InStock
102994-468
H pylori Antigens ELISA Assay Kit (Quantitative), Eagle Biosciences, Inc.
Assays ELISAs
Helicobacter pylori Antigen ELISA determines H
  • Dynamic Range: 1.9 - 150 ng/ml
  • Sensitivity: 0.165 ng/ml
  • Incubation: 2 hours
H. pylori (previously known as Campylobacter pyloridis) is a type of bacteria that infects the stomach and is a common cause of peptic ulcers. H. pylori bacteria can be passed from person to person through direct contact with saliva, vomit or fecal matter. H. pylori can also be spread through contaminated food or water.
pylori antigen in fecal samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty