Would you like to log in again?
Anti-Human Tissue Transglutaminase (anti-tTG) IgA ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4033
102994-470EA 797.69 USD InStock
102994-470
Anti-Human Tissue Transglutaminase (anti-tTG) IgA ELISA, Eagle Biosciences
Assays ELISAs
Anti-Human tTG ELISA determines IgA autoantibodies against human tissue transglutaminase in serum.
  • Dynamic Range: 10 - 300 U/ml
  • Sensitivity: 3 U/ml
  • Incubation: 2 hour
The Eagle Biosciences Anti-Human Tissue Transglutaminase (anti-tTG) ELISA Assay Kit is designed and validated for the quantitative determination of IgA autoantibodies against human tissue transglutaminase in human serum. The Anti-Human Tissue Transglutaminase (anti-tTG) ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty