Would you like to log in again?
IAA (Insulin Autoantibodies) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
3806
102994-476EA 778.08 USD InStock
102994-476
IAA (Insulin Autoantibodies) ELISA, Eagle Biosciences
Assays ELISAs
IAA ELISA is an immuno to detect IgG autoantibodies and antibodies to insulin in human serum.
  • Dynamic Range: 0.1 - 20 U/mL
  • Sensitivity: 0.08 U/mL
  • Incubation: 90 minutes
Type 1 diabetes, also known as insulin-dependent diabetes mellitus (IDDM), results from a chronic autoimmune destruction of the insulin-secreting pancreatic beta cells, probably initiated by exposure of genetically susceptible host to an environmental agent. Autoimmune destruction of beta cells is thought to be completely asymptomatic until 80-90% of the cells are lost. This process may take years to complete and may occur at any time in all ages.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty