Would you like to log in again?
Urinary Isoprostane ELISA Kit, Eagle Biosciences
Supplier: Eagle Biosciences
ISO34-K01
102994-570EA 712.69 USD InStock
102994-570
Urinary Isoprostane ELISA Kit, Eagle Biosciences
Assays ELISAs
Urinary Isoprostane ELISA is quantitative determination of Isoprostane in human urine by microplate.
  • Dynamic Range: 0.05 - 100 ng/mL
  • Sensitivity: 0.05 ng/mL
  • Incubation: 2.5 hours
Isoprostanes are prostaglandin-like compounds that are produced by free radical mediated peroxidation of lipoproteins. This kit is for the quantification of 15-isoprostane F2t (also known as 8-epi-PGF2a or 8-iso-PGF2a) in urine samples. Levels of 15-isoprostane F2t in urine are useful for the non-invasive assessment of oxidant stress in vivo. 15-isoprostane F2t has also been shown to be a potent vasoconstrictor in rat kidneys and rabbit lungs, and plays a causative role in atherogenesis. Elevated isoprostane levels are associated with hepatorenal syndrome, rheumatoid arthritis, atherosclerosis, and carcinogenesis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty