Would you like to log in again?
DM1 Antibody Drug Conjugate (ADC) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
KTR-756
102994-678EA 2909.62 USD InStock
102994-678
DM1 Antibody Drug Conjugate (ADC) ELISA, Eagle Biosciences
Assays ELISAs
DM1 Antibody Drug Conjugate (ADC) ELISA determines antibody-DM1-conjugate level in test samples.
  • Dynamic Range: 0.016 - 1.0 µg/mL
  • Sensitivity: 0.0077 µg/mL
  • Incubation: 2.5 hours
The Eagle Biosciences DM1 Antibody Drug Conjugate (ADC) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for use in the quantitative determination of antibody-DM1-conjugate (Trastuzumab emtansine) level in test samples. It is useful for pre-clinical and clinical pharmacology study of DM1 Antibody Drug Conjugate (ADC).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty