Would you like to log in again?
Human IL-1β ELISA KIT, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
L1B31-K01
102994-720EA 660.38 USD InStock
102994-720
Human IL-1β ELISA KIT, Eagle Biosciences, Inc.
Assays ELISAs
Human Interleukin-1 Beta ELISA detects IL-1 Beta concentrations in biological samples.
  • Dynamic Range: 7.8 - 250 pg/ml
  • Sensitivity: 2 pg/ml
  • Incubation: 3.5 hours
Interleukin-1 (IL-1), originally described as lymphocyte activating factor (LAF) for its effects on thymocytes (1), is a polypeptide cytokine with two molecular forms. The two distinct molecular forms of IL-1 are thought to be derived from two genes (2,3). After transcription, as 31kD precursor polypeptide is cleaved to give rise to mostly cell membrane associated IL-1a and secreted IL-1b. Both have the same molecular weight of 15kD but have different isoelectric points of 5 and 7, respectively.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty