Would you like to log in again?
Mouse IL-25 (IL-17E) ELISA KIT, Eagle Biosciences
Supplier: Eagle Biosciences
L2511-K01
102994-732EA 660.38 USD InStock
102994-732
Mouse IL-25 (IL-17E) ELISA KIT, Eagle Biosciences
Assays ELISAs
Mouse IL-25 IL-17E EIA detects mouse IL-25, IL-17E concentrations in cell culture supernates, serum.
  • Dynamic Range: 31.25 - 1000 pg/ml
  • Sensitivity: 15 pg/ml
  • Incubation: 3.5 hours
IL-25 (also known as IL-17E) is a member of the IL-17 family of proinflammatory cytokines, which includes IL-17A, IL-17B, IL-17C, IL-17F and IL-17A/F. IL-17E is found at low levels in various peripheral tissues. It is a proinflammatory cytokine that promotes airway eosinophilia in mice. IL-17E plays a role in Th2 type inflammatory diseases, such as asthma and atopic dermatitis. IL-25 induces elevated gene expression of IL-4, IL-5 and IL-13 in multiple tissues and results in T helper 2 (TH2)-type immune responses (increased serum IgE levels) and pathologic changes in the lungs and digestive tract with eosinophilic infiltrates, increased mucus production, epithelial cell hyperplasia and overall amplified allergic inflammation. IL-25 shares the receptor IL-17RB with IL-17B, although it binds with much higher affinity than IL-17B.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty