Would you like to log in again?
Lipoxin A4 ELISA Kit, Eagle Biosciences
Supplier: Eagle Biosciences
LA439-K01
102994-744EA 1209.62 USD InStock
102994-744
Lipoxin A4 ELISA Kit, Eagle Biosciences
Assays ELISAs
Lipoxin A4 ELISA is to detect Lipoxin A4 in biological fluids by a microplate enzyme immuno.
  • Dynamic Range: 0.02 - 2.0 ng/mL
  • Sensitivity: 0.02 ng/mL
  • Incubation: 2 hours
Lipoxin A4 (LXA4) is a biologically active lipoxygenase interaction product derived from arachidonic acid. Arachidonic acid is first oxygenated by 15-lipoxygenase to form 15-HETE which is converted by 5-lipoxygenase and epoxide hydrase to generate LXA4. LXA4 stimulates leukocyte chemotaxis without aggregation and inhibits natural killer cell cytotoxicity. It also provokes contraction of parenchymal strips and stimulates microvascular changes. Recent findings indicate that LXA4 inhibits leukocyte-dependent inflammation. Determination of LXA4 level may provide new understanding of the role of LXA4 in basic cellular reactions and in pathophysiology of inflammation and other disease processes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty