Would you like to log in again?
LPO Microplate Based Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
LIP39-K01
102994-760EA 568.85 USD InStock
102994-760
LPO Microplate Based Assay Kit, Eagle Biosciences
Assays ELISAs
Lipid Peroxidase detects malondialdehyde (MDA) or MDA in combo with 4-HAE in biological fluids.
  • Dynamic Range: 0.5 - 15 µM
  • Sensitivity: 0.1 nmol/mL
  • Incubation: 3 hours
Lipid peroxidation is a well-established mechanism of cellular injury in both plants and animals, and is used as an indicator of oxidative stress in cells and tissues. Lipid peroxides are unstable and decompose to form a complex series of compounds including reactive carbonyl compounds. Polyunsaturated fatty acid peroxides generate malondialdehyde (MDA) and 4-hydroxyalkenals (HAE) upon decomposition. The measurement of MDA and HAE has been used as an indicator of lipid peroxidation (1). This method is designed to assay either MDA alone (in hydrochloric acid) or MDA in combination with HAE (in methanesulfonic acid.) With this kit, 40 samples can be assayed in duplicate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty