Would you like to log in again?
Human MCP-1 ELISA KIT, Eagle Biosciences
Supplier: Eagle Biosciences
MCP31-K01
102994-778EA 660.38 USD InStock
102994-778
Human MCP-1 ELISA KIT, Eagle Biosciences
Assays ELISAs
Human Monocyte Chemoattractant Protein-1 ELISA determines MCP-1 concentrations in various samples.
  • Dynamic Range: 31.25 - 2000 pg/ml
  • Sensitivity: 15 pg/ml
  • Incubation: 3.5 hours
Monocyte chemoattractant protein-1 (MCP-1) called also monocyte chemotactic protein-1. MCP-1 is an 8.6kDa protein and belongs to the family of chemotactic cytokines known as Chemokines. MCP-1 is expressed by monocytes, vascular endothelial cells, smooth muscle cells, glomerular mesangial cell, osteoblastic cells, and human pulmonary type-2-like epithelial cells in culture. MCP-1 exhibits chemotactic activity for monocytes/macrophages, basophils, T lymphocytes, particularly memory T cells and natural killer cells and neural stem cells. Elevated levels of MCP-1 are detected during inflammation and immune responses. MCP-1 activity is also important in wound healing as shown by delayed wound repair in MCP-1deficient mice. MCP-1 is implicated in the pathogenesis of disease states that involve monocyte/macrophage infiltrates, such as psoriasis, rheumatoid arthritis and recruitment of monocytes to the arterial wall in atherosclerosis. In addition to its chemotactic function, MCP-1 also induces the expression of IL-10 from macrophages, which favors a TH2 immune response. MCP-1 is a glycopeptide containing one potential N-linked glycosylation site. The extent of MCP-1 glycosylation influences its chemotactic potency and half-life in vivo.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty