Would you like to log in again?
1-Methylhistamine Urine ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
EA208/96
102994-784EA 836.92 USD InStock
102994-784
1-Methylhistamine Urine ELISA, Eagle Biosciences, Inc.
Assays ELISAs
1-Methylhistamine ELISA detects 1-Methylhistamine in biological samples.
  • Dynamic Range: 10 - 1000 ng/ml
  • Sensitivity: 0.13 ng/ml
  • Incubation: overnight
The Eagle Biosciences 1-Methylhistamine ELISA Assay kit is an Enzyme-linked immunosorbent assay used for the quantitative and very sensitive determination of noradrenaline in biological samples including urine, plasma, and cell culture samples. The 1-Methylhistamine ELISA Assay kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty