Would you like to log in again?
Rat MIP-2 ELISA KIT, Eagle Biosciences
Supplier: Eagle Biosciences
MIP21-K01
102994-790EA 660.38 USD InStock
102994-790
Rat MIP-2 ELISA KIT, Eagle Biosciences
Assays ELISAs
Rat MIP-2 ELISA detects rat macrophage inflammatory protein-2 concentration in various sample types.
  • Dynamic Range: 62.5 - 2000 pg/ml
  • Sensitivity: 15 pg/mL
  • Incubation: 3.5 hours
The Eagle Biosciences Rat Macrophage inflammatory protein-2 (MIP-2) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of rat macrophage inflammatory protein-2 (MIP-2 ) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Rat Macrophage inflammatory protein-2 (MIP-2) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty