Would you like to log in again?
MMP-7 ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
MM731-K01
102994-792EA 1340.38 USD InStock
102994-792
MMP-7 ELISA, Eagle Biosciences
Assays ELISAs
MMP-7 ELISA is intended to detect the Cancer Antigen MMP-7 concentration in human serum.
  • Dynamic Range: 1 - 256 ng/ml
  • Sensitivity: 1 ng/ml
  • Incubation: 3 hours
MMP-7 proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The MMP-7 enzyme degrades proteoglycans, fibronectin, elastin and casein and differs from most MMP family members in that it lacks a conserved C-terminal protein domain. The enzyme is involved in wound healing, and studies in mice suggest that it regulates the activity of defensins in intestinal mucosa. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty