Would you like to log in again?
Myeloperoxidase (MPO) Stool, Urine ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
MPO39-K01
102994-798EA 882.69 USD InStock
102994-798
Myeloperoxidase (MPO) Stool, Urine ELISA, Eagle Biosciences
Assays ELISAs
Myeloperoxidase (MPO) ELISA determines human MPO levels in stool and urine samples.
  • Dynamic Range: 2 - 512 ng/ml
  • Sensitivity: 0.3 ng/ml
  • Incubation: 2.5 hours
Myeloperoxidase (MPO) is a specific polymorphonuclear enzyme that is most abundantly expressed in neutrophil granulocytes. It functions in the oxygen-dependent killing of microorganisms, and is released from primary granules of neutrophils during acute inflammation. MPO is the product of a single gene, which is about 11 kb in size, composed of 11 introns and 12 exons, and located in the long arm of chromosome 17 in segment q12-24. The mature 150 kDa MPO protein is a dimer consisting of two 15 kDa light chains and two heavy chains of variable degrees of glycosylation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty