Would you like to log in again?
NGAL (Stool) ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
NGL35-K01
102994-806EA 765 USD InStock
102994-806
NGAL (Stool) ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
NGAL (Stool) ELISA detects human neutrophil gelatinase-associated lipocalin in feces.
  • Dynamic Range: 1.2 - 97.2 µg/g
  • Sensitivity: 0.04 µg/g
  • Incubation: 2 hours
NGAL or neutrophil gelatinase-associated lipocalin also known as Lipocalin-2 (LCN2) or oncogene 24p3 is a protein, which in humans is encoded by the LCN2 gene.[1][2][3] NGAL is involved in innate immunity by sequestrating iron that in turn limits bacterial growth.[4] It is expressed in neutrophils and in low levels in the kidney, prostate, and epithelia of the respiratory and alimentary tracts.[3][5]Studies have shown that NGAL is an early biomarker for ischaemic renal injury after cardiopulmonary bypass.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty