Would you like to log in again?
Normetanephrine and Metanephrine (Plasma) ELISA Kit, Eagle Biosciences
Supplier: Eagle Biosciences
EA612/192
102994-810EA 836.92 USD InStock
102994-810
Normetanephrine and Metanephrine (Plasma) ELISA Kit, Eagle Biosciences
Assays ELISAs
Normetanephrine and Metanephrine ELISA detects normetanephrine and metanephrine in plasma.
  • Dynamic Range: Metanephrine: 20 - 3000 pg/ml, Normetanephrine: 20 - 3000 pg/ml
  • Sensitivity: Metanephrine: 10 pg/ml, Normetanephrine: 15 pg/ml
  • Incubation: Overnight
Normetanephrine and metanephrine are physiologically formed from the catechol-amines noradrenaline and adrenaline by the enzyme catechol-O-methyltransferase (COMT). Increased levels of normetanephrine and metanephrine can be found in pheochromocytoma, ganglioneuroma and other neurogenic tumors. The Normetanephrine & Metanephrine (Plasma) ELISA assay kit provides materials for the quantitative measurement of free normetanephrine and metanephrine in EDTA plasma. Normetanephrine and metanephrine are quantitatively acylated to their N-acyl-derivates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty