Would you like to log in again?
Noradrenaline (Norepinephrine) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
EA610/96
102994-814EA 738.85 USD InStock
102994-814
Noradrenaline (Norepinephrine) ELISA, Eagle Biosciences
Assays ELISAs
Noradrenaline (Norepinephrine) ELISA detects Noradrenaline in urine or plasma by a microplate.
  • Dynamic Range: 1.5 - 500 ng/ml
  • Sensitivity: Urine: 0.85 ng/ml, Plasma: 43 pg/ml
  • Incubation: Overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitters, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty