Would you like to log in again?
Colorimetric Nitric Oxide Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
NSC31-K01
102994-820EA 529.62 USD InStock
102994-820
Colorimetric Nitric Oxide Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Nitric Oxide Synthase Colorimetricdetermines Nitric Oxide Synthase in biological fluids.
  • Dynamic Range: 0.5 - 100 µM
  • Sensitivity: 1 pmol/µL or ~1 µM
  • Incubation: 5 minutes
The Nitric Oxide Synthase Colorimetric Assay kit employs immunoaffinity purified Nitrate Reductase (NaR) enabling the measure of total nitric oxide (NO) produced in in vitro experimental systems. Nitric oxide can be spectrophotometrically assayed by measuring the accumulation of its stable degradation products, nitrate and nitrite. The ratio of these two products in biological fluids, tissue culture media, etc. may vary substantially. Hence, for accurate assessment of the total nitric oxide generated, one must monitor both nitrate and nitrite. An excellent solution to this problem is the enzymatic conversion of nitrate to nitrite by the enzyme nitrate reductase, followed by quantitation of nitrite using Griess Reagent. In addition to providing all necessary components in a microtiter format, this kit employs affinity purified nitrate reductase and NADH, thereby circumventing some of the potential problems reported for NO measurement using NADPH dependent nitrate reductases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty