Would you like to log in again?
Osteocalcin ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
OST31-K01
102994-826EA 765 USD InStock
102994-826
Osteocalcin ELISA, Eagle Biosciences
Assays ELISAs
Osteocalcin ELISA detects human osteocalcin (1-49) and osteocalcin (1-43) levels.
  • Dynamic Range: 4.0 - 64 ng/ml
  • Sensitivity: .31 ng/ml
  • Incubation: 1.5 hours
The Eagle Biosciences Human Osteocalcin ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of both human osteocalcin (1-49) and osteocalcin (1-43) (also referred as N-terminal & mid-regional osteocalcin) levels in test samples). The Eagle Biosciences Human Osteocalcin ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty