Would you like to log in again?
Potential Antioxidant Capacity Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
PAC39-K01
102994-838EA 2059.62 USD InStock
102994-838
Potential Antioxidant Capacity Assay, Eagle Biosciences, Inc.
Assays ELISAs
Antioxidant Capacity Potential detects of adduct antioxidant capacity in biological samples.
  • Dynamic Range: 21.9 - 4378 µmol/L (cupric ion reducing power)
  • Sensitivity: 0.063 mM
  • Incubation: 3 minutes
Oxidative stress plays on important role in various diseases and aging. The control of oxidative stress is expected to be useful to prevent diseases and aging. Oxidative stress is caused by the imbalance between reactive oxygen species (ROS) and antioxidant defense system. For accurate assessment of oxidative stress, measurement of ROS, oxidative damage and antioxidant activity may be essential. The Eagle Biosciences Potential Antioxidant Capacity Assay can detect not only hydrophilic antioxidants such as Vitamin C and glutathione, but also can detect hydrophobic antioxidants such as Vitamin E. This Antioxidant Capacity Potential Assay kit is applicable for assessment of total antioxidants of serum, foods and beverage samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty