Would you like to log in again?
Anti-Phosphatidylserine ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4056
102994-842EA 778.08 USD InStock
102994-842
Anti-Phosphatidylserine ELISA, Eagle Biosciences
Assays ELISAs
Anti-Phosphatidylserine ELISA detects IgG and IgM Ab to phosphatidylserine in human serum.
  • Dynamic Range: 3 -300 U/ml
  • Sensitivity: 3 U/ml
  • Incubation: 2.5 hours
APAS is an autoimmune disorder comprising such clinical symptoms like arterial or venous thrombosis, thrombocytopenia and recurrent fetal loss. Primary APAS as well as systemic lupus erythematosus (SLE) are characterized by the appearance of autoantibodies to negatively charged phospholipids (1). Although significance and pathological relevance of phospholipid antibodies are not completely revealed yet, the detection of such autoantibodies is widely established and plays an important role in the diagnostics of systemic autoimmune diseases.  Unlike phospholipid antibodies which appear in some infectious disease patients autoimmune patients exhibit phospholipid antibodies that seem to recognize phospholipids in association with plasma protein cofactors such as ß2-glycoprotein I (ß2-GP I, apolipoprotein H) (2). ß2-GP I, a serum protein with a molecular weight of 50 kDa, affects platelet aggregation and coagulation. The positively charged fifth domain of ß2-GP I interacts with negatively charged phospholipids such as phosphatidyl-serine. This interaction results in conformational changes of the protein and the creation of new epitopes apparently recognized by autoimmune phospholipid autoantibodies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty