Would you like to log in again?
Human Perforin ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
PFN31-K01
102994-848EA 830.38 USD InStock
102994-848
Human Perforin ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human Perforin ELISA to determine Perforin in supernatant, buffered solutions.
  • Dynamic Range: 62.5-2000 pg/ml
  • Sensitivity: 40 pg/ml
  • Incubation: 2h45
The Eagle Biosciences Human Perforin ELISA Assay Kit is to be used for the in-vitro quantitative determination of Perforin in supernatant, buffered solutions. The assay will recognize natural Perforin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty