Would you like to log in again?
Phenylalanine, Tyrosine & Tryptophan HPLC Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
PNL31-H100
102994-886EA 1006.92 USD InStock
102994-886
Phenylalanine, Tyrosine & Tryptophan HPLC Assay, Eagle Biosciences, Inc.
Assays Immunological Assays
The Phenylalanine Tyrosine & Tryptophan HPLC Assay detects phenylalanine, tyrosine and tryptophan.
  • Incubation Time: 15 minutes
  • Sample Size: 50 µl
  • Sample Type: plasma, whole blood, filter spots
The Eagle Biosciences Phenylalanine, Tyrosine & Tryptophan HPLC Assay kit makes it possible to determine phenylalanine, tyrosine and tryptophan in an easy, fast and precise method. The Phenylalanine, Tyrosine & Tryptophan HPLC Assay kit includes all reagents for preparation and separation of the samples with exception of the columns (IC4000rp) and the controls (IC4000ko). Both can be supplied by Eagle Biosciences. Besides the complete test kits it is possible to order all components separately. Please request our single component price list.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty