Would you like to log in again?
Pepsinogen II ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
PP231-K01
102994-890EA 810.77 USD InStock
102994-890
Pepsinogen II ELISA, Eagle Biosciences
Assays ELISAs
Human Pepsinogen II ELISA is intended to detect human pepsinogen II levels in serum.
  • Dynamic Range: 6.3 - 100 ng/ml
  • Sensitivity: 0.1 ng/ml
  • Incubation: 2.5 hours
The Eagle Biosciences Human Pepsinogen II ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human pepsinogen II levels in serum. The Eagle Biosciences Human Pepsinogen ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty