Would you like to log in again?
Porphyrins HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
PPR34-H100
102994-892EA 1006.92 USD InStock
102994-892
Porphyrins HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Porphyrins HPLC Assay Kit is intended to determine porphyrins in the urine.
  • Incubation Time: 5 minutes
  • Sample Size: 1 mL
  • Sample Type: Urine
Hemoglobin, myoglobin and the cytochromes contain the heme structure as active site. The porphyrins are precursors of the heme molecules. They play an important role in the oxygen metabolism. The heme synthesis is located mainly in the erythrocyte cells. Glycin and succinyl-CoA are condensed to δ-aminolävulinic acid. Two molecules of δ-aminolävulinic are then condensed to porphobilinogen. The porphyrin ring is built up by four porphobilinogen molecules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty