Would you like to log in again?
Rat Prorenin/Renin Total ELISA Kit, Eagle Biosciences
Supplier: Eagle Biosciences
PRO29-K01
102994-904EA 1412.31 USD InStock
102994-904
Rat Prorenin/Renin Total ELISA Kit, Eagle Biosciences
Assays ELISAs
Rat Prorenin Renin ELISA determines Rat Prorenin and Renin level in biological fluids by ELISA.
  • Dynamic Range: 0.1 - 100 ng/mL
  • Sensitivity: 0.1 ng/mL
  • Incubation: 1.5 hours
Prorenin is a glycosylated aspartic protease that consists of 2 homologous lobes and is the precursor of renin. Renin activates the renin-angiotensin system by cleaving angiotensinogen, produced by the liver, to yield angiotensin I, which is further converted into angiotensin II by ACE, the angiotensin-converting enzyme primarily within the capillaries of the lungs. It has been reported that the levels of circulating prorenin (but not renin) are increased in diabetic subjects
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty