Would you like to log in again?
RF IgM (Rheumatoid Factor) ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
4046
102994-912EA 568.85 USD InStock
102994-912
RF IgM (Rheumatoid Factor) ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Rheumatoid Factor IgM ELISA is used to detect rheumatoid factor (RF) IgM in human serum or plasma.
  • Dynamic Range: 1-300 U/mL
  • Sensitivity: 1U/mL
  • Incubation: 2.5 hours
Patients suffering from rheumatoid arthritis (RA) exhibit RF, autoantibodies recognizing the Fc part of IgG. RA or chronic polyarthritis has a yet unknown etiology and represents the most frequent rheumatic inflammatory disorder demonstrating a prevalence rate of up to 1%. One of the typical manifestations of RA is symmetric synovialitis of limb joints often accompanied by involvement of the cervical spinal column. Beside clinical features one of the criteria of the American College of Rheumatology for the diagnosis of RA is the presence of RF (1). Up to 80 % of RA patients may demonstrate RF. RF can occur years prior to the onset of disease and RF positive apparently healthy individuals bear a 5 - 40 times higher risk to develop RA (2). However, patients suffering from other autoimmune, infectious or B-cell lympho¬pro¬liferative disorders as well as apparently healthy elderly individuals may develop RF.High concentrations of RF are often associated with a more severe disease comprising a faster destruction of joints. In addition, they are found in patients with extra-articular manifestations such as rheumatoid nodules, polyneuropathy, vasculitis or Sicca syndrome.RF may belong to the IgG, IgM or IgA isotype whereas IgM RF is the most frequent isotype to be determined in RA patients. Extra-articular manifestations seem to be associated with IgA RF. Like RF of the IgM isotype high concentrations of IgG RF seems to appear with patients suffering from more progressive erosions of joint
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty