Would you like to log in again?
Serotonin ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
EA602/96
102994-986EA 640.77 USD InStock
102994-986
Serotonin ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Serotonin ELISA detects the concentration of serotonin in Serum, Plasma, and Urine.
  • Dynamic Range: 15 - 2500 ng/mL
  • Sensitivity: 5 ng/mL
  • Incubation: 2.5 hours
Serotonin (5-Hydroxytryptamine), a biogenic amine, is a product of the tryptophan metabolism. It is a well evaluated neurotransmitter of the central nervous system and can be found in high concentrations in the chromaffine cells of the intestinal mucosa, in the platelets and the serotonergic neurones of the brain. Central-serotonergic neurons influence physiological functions such as sleep and the hormonal and cardio-vascular regulation. Increased serum levels can be found with malignant carcinoid, endogenous depression and schizophrenia.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty