Would you like to log in again?
Secretory Leukocyte Peptidase Inhibitor Protein (SLPI) ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
SLP31-K01
102994-994EA 1340.38 USD InStock
102994-994
Secretory Leukocyte Peptidase Inhibitor Protein (SLPI) ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Secretory Leukocyte Peptidase Inhibitor Protein ELISA detects SLPI concentration in human serum.
  • Dynamic Range: 1 - 256 ng/ml
  • Sensitivity: 1 ng/ml
  • Incubation: 2.5 hours
The Secretory Leukocyte Peptidase Inhibitor Protein (SLPI) ELISA assay kit is a secreted inhibitor which protects epithelial tissues from serine proteases. It is found in various secretions including seminal plasma, cervical mucus, and bronchial secretions, and has affinity for trypsin, leukocyte elastase, and cathepsin G. Its inhibitory effect contributes to the immune response by protecting epithelial surfaces from attack by endogenous proteolytic enzymes. The SLPI (ALP) protein is also thought to have broad-spectrum antibiotic activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty