Would you like to log in again?
Total Antioxidant Power Microplate Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
TAP39-K01
102995-006EA 699.62 USD InStock
102995-006
Total Antioxidant Power Microplate Assay Kit, Eagle Biosciences
Assays ELISAs
Total Antioxidant Power Microplate detects the total anti-oxidative power in biological fluids.
  • Dynamic Range: 0.125 - 2.0 mM
  • Sensitivity: 0.125 ng/mL
  • Incubation: 3 minutes
Oxidative stress has been implicated in a number of diseases such as atherosclerosis1, chronic inflammatory disease2, chronic renal failure3, and cancer4. It is a condition where an imbalance exists between the production of reactive oxidizing species and the body’s ability to neutralize these intermediates, resulting in cellular damage. The body has designed several physiological responses to oxidative stress including counterbalances5 such as enzymes and variously functionalized molecules (see examples below) that effectively neutralize these damaging species. These antioxidants can be either water or lipid soluble, and are localized transiently throughout various tissues, cells and cell types.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty